Learn More
Strem, An Ascensus Company CAS# 7782-61-8. 5kg. Iron(III) nitrate nonahydrate, 98+%. MFCD00149708
SDP
Supplier:  Strem, An Ascensus Company 9326145000G
Encompass
CAS# 7782-61-8. 5kg. Iron(III) nitrate nonahydrate, 98+%. MFCD00149708. Molecular Weight: 241.86 (404.02). Molecular Formula: Fe(NO3)3∙9H2O. Color/form: light purple xtl. Strem# 93-2614. http://www.strem.com/catalog/v/93-2614/
Catalog No. 50-901-14640