Learn More
Sigma Aldrich Fine Chemicals Biosciences Iron oxideIIIII magnetic n
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 725358
Encompass
Concentration 5mg/ml includes total weight nanocrystals plus ligands.
Catalog No. 50-290-6937