Learn More
Perkin Elmer US LLC Iron Reference Material - Wire, 300 mg, 780°C
Supplier:  Perkin Elmer US LLC 09988017
Encompass_Preferred
PerkinElmers Calibration and Reference Standards ensure accurate thermogravimetric measurements for all of PerkinElmers TGA instruments. Magnetic calibration and reference standards are available as a kit, as well as individually. PerkinElmer recommends the use of a fresh reference material every time you calibrate your instruments. This helps to avoid the effects of hysteresis and oxidation on the Curie Point of the reference materials.
Catalog No. 50-211-2706