Learn More
Hach Company Iron Test Kit Model IR-10
Supplier:  Hach Company 146430
Encompass
Includes one reagent-two color discs-comparator box-two viewing tubes-instructions and carrying case Using the simple color disc method this kit measures Iron (medium range 0 1 - 10 mg/L) and contains reagent for 100 tests
Catalog No. 50-002-57152