Learn More
Ironclad Performance Wear Ironclad Ranchworx Glove, 12 pr per box, size XXXLarge, Black/Tan, EN388
Supplier:  Ironclad Performance Wear RWG207XXXL
Encompass_Preferred
Gloves, Ironclad, Ranchworx Glove,Premier leather work glove, Roll-top fingertip design, Grain goat leather padded palm, Back of and TPR impact protection, Thumb nail guard. Ideal for Ranching, Forestry, HVAC, Heavy cable and wiring. 12 pr per box.
Catalog No. 50-161-0050