Learn More
Sigma Aldrich Fine Chemicals Biosciences IronII chloride tetrahydra50G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 4493950G
Encompass
Iron(II) chloride tetrahydrate is a hydrated metallic halide. Crystal structure studies suggest that it crystallizes in monoclinic system having space group P21/c.
Catalog No. 50-289-8655