Learn More
Sigma Aldrich Fine Chemicals Biosciences IronII sulfate heptahydrat5KG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 12354
Encompass
Iron(II) sulfate heptahydrate is a heptahydrate of iron(II) sulfate. It undergoes thermal degradation to afford hydrosulfate and oxysulfate of iron(III). Isothermal and dynamic thermal analysis methods have been proposed to study its thermal decomposition in air.
Catalog No. 50-290-6674