Learn More
FUJIFILM BIOSCIENCES INC IronII Sulfate Heptahydrate
Supplier:  FUJIFILM BIOSCIENCES INC 09501095
Encompass_Preferred
Pale bluish green crystals freely soluble in water practically insoluble in ethanol For research use only RUO
Catalog No. 50-004-50233