Learn More
Lonza Walkersville Irrad Mouse Embryonic Fibrobl
Supplier:  Lonza Walkersville MIFB482
Encompass
Irradiated Mouse primary embryonic fibroblasts (iMEF) are dissociated from day 14 and 15 CD1 mouse embryos expanded and then inactivated using gamma radiation Each vial contains at least 2 million cells iMEF are convenient and easy to use ldquo feeder cells rdquo commonly co-cultured with embryonic and pluripotent stem cell cultures Recommended Media Lonza s DMEM High Glucose w/ L-Gln 500 mL (Part 12-604F) containing 10% FBS (Sigma-Aldrich Part F6178)
Catalog No. 50-004-96351