Learn More
Med Vet International Irrigating Cannula Small, J0150A, 5 Pack
Supplier:  Med Vet International JOR150A
Encompass_Preferred
These plastic cannula are used for irrigation of the lacrimal ducts, wounds, glands, etc. Rounded ends. Flexible tip. Sterile.
Catalog No. 50-005-83711