Learn More
Enzo Life Sciences IRS-1 (Tyr608) peptide (1mg)
SDP
Supplier:  Enzo Life Sciences BMLP3200001
Encompass_Preferred
This sequence, KKHTDDGYMPMSPGVA, is from insulin receptor substrate-1 (IRS-1) inclusive of Tyr608 (mouse) -Tyr612 (human). It contains the insulin receptor tyrosine kinase substrate motif YMXM (Tyr-Met-X-Met). This peptide has been used as a substrate for purified insulin receptor (Km=90 µM) and other tyrosine kinases in phosphocellulose binding assays. The tyrosine phosphorylated version of this peptide binds to phosphatidylinositol 3-kinase (PI 3-kinase) SH2 domain and activates the enzyme. Alternative name: Insulin receptor substrate (Tyr608) peptide. Purity: ≥95% (HPLC). Formulation: Lyophilized. Solubility: Soluble in water. Long Term Storage: -20°C.
Catalog No. 50-201-1344