Learn More
Medchemexpress LLC HY-16937 100mg , ISA-2011B CAS:1395347-24-6 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY16937100MG
Encompass_Preferred
Medchemexpress, HY-16937 100mg ISA-2011B CAS:1395347-24-6 Formula:C22H18ClN3O4 Purity:98% ISA-2011B is a PIP5Kα inhibitor with promising anticancer effects . Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2719