Learn More
Apexbio Technology LLC Isatin 91-56-5 500mg
SDP
Supplier:  Apexbio Technology LLC B6153500
Encompass_Preferred
Isatin (CAS 91-56-5) is a small molecule inhibitor of monoamine oxidase (MAO) exhibiting an IC50 of 15 M Monoamine oxidase inhibitors are investigated for their roles in modulating neurotransmitter metabolism and have been explored in the management of depressive disorders and neurodegenerative diseases such as Parkinson s disease In vitro exposure of SH-SY5Y neuroblastoma cells to isatin induces a dose-dependent increase in MTT metabolism and at higher concentrations promotes cell death characterized by a shift from apoptosis at lower doses to increased necrosis at higher doses In vivo isatin treatment in a rat model of Japanese encephalitis virus-induced Parkinsonism enhances motor activity and prevents dopamine depletion in the striatum indicating its utility in neuropharmacological research
Catalog No. 50-000-89654