Learn More
Medchemexpress LLC Isavuconazole | 241479-67-4 | 99.9% | C22H17F2N5OS | 200 MG
SDP
Supplier:  Medchemexpress LLC HY14273200MG
Encompass_Preferred
Isavuconazole is a triazole proagent that exhibits antifungal activity against yeasts, molds, and dimorphic fungi. It functions by inhibiting ergosterol biosynthesis, which leads to the disruption of the fungal membrane structure and function. Isavuconazole is also identified as a moderate inhibitor of CYP3A4.
  • Triazole proagent
  • Exhibits antifungal activity against yeasts, molds, and dimorphic fungi
  • Inhibits ergosterol biosynthesis
  • Disrupts fungal membrane structure and function
  • Moderate inhibitor of CYP3A4
Catalog No. 50-003-53500