Learn More
TARGETMOL CHEMICALS INC ISAVUCONAZOLE 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T230525MG
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 10 mg 50 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Isavuconazole (RO-0094815)(BAL-4815; RO-0094815) is the active component of the new azole antifungal agent BAL8557 with MIC(50)s/MIC(90)s ranged from 0.002/0.004 mg/liter for C. albicans to 0.25/0.5 mg/liter for C. glabrata. purity: 99%
Catalog No. 50-258-0898