Learn More
STEMCELL Technologies ISCK03, Size: 10 mg
SDP
Supplier:  STEMCELL Technologies 73734
Encompass
ISCK03 (inhibitor of SCF/c-KIT) is a cell-permeable phenyl-imidazolosulfonamide compound that inhibits KIT activity. ISCK03 inhibits SCF-induced c-KIT phosphorylation and downstream ERK phosphorylation at a concentration of 1 - 5 µM in 501mel melanoma cells (Na et al.).CANCER RESEARCH· Causes significant decrease in blood vessel density on human lung adenocarcinoma (A549) cells irradiated with X-rays (6 Gy; Kamlah et al.).· Reduces cell viability and ERK1/2 phosphorylation in acute myeloid leukemia (AML)-derived cell lines (Kon et al.).
Catalog No. 50-197-6122
Add to Cart
Description
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov