Learn More
Bioworld Iscove's DMEM w/o Phenol Red, 500 mL
Supplier:  Bioworld 306110471
Encompass_Preferred
Guilbert and Iscove published their modifications to Dulbecco's Modified Eagle's Medium (DMEM) in 1976. Their modification included the supplements of selenium, additional vitamins and amino acids, sodium pyruvate and HEPES buffer. In addition, ferric nitrate was replaced with potassium nitrate. When further supplemented with albumin, lecithin and transferrin, this medium supported the growth of precursor cell of macrophages and erythocytes without serum supplementation. Iscove's Medium has since been shown to support murine B lymphocytes, hemopoietic tissue from bone marrow, B cells stimulated with lipopolysaccharide, T lymphocytes and a variety of hybrid cells. Iscove's Modified Dulbecco's Media (DMEM) liquid, item #785100 , contains l-glutamine and 25mM HEPES buffer. Does not contain phenol red. Click here to see an Iscove's Modified Dulbecco's Medium Components Table.
Catalog No. 50-254-114