Learn More
Medchemexpress LLC Iseganan | 257277-05-7 | 98.9% | 1900.28 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP35125MG
Encompass_Preferred
Iseganan is an anti-microbial peptide active against Gram-positive and Gram-negative bacteria and fungi. It kills a broad spectrum of bacteria and fungi by attaching to and destroying the integrity of lipid cell membranes. This peptide can be used for oral mucositis research.
  • Active against Gram-positive and Gram-negative bacteria and fungi
  • Disrupts T. crassiceps cysticerci in vitro
  • Shows anti-parasite activity in T. crassiceps murine cysticercosis model
Catalog No. 50-003-97395