Learn More
Medchemexpress LLC Isepamicin sulfate | 67814-76-0 | 98.0% | 569.6 g/mol | C22H43N5O12·xH2SO4 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY100589100MG
Encompass_Preferred
Isepamicin sulfate is a broad-spectrum aminoglycoside antibiotic supplied as the sulfate salt for research use. It exhibits activity against certain Gram-negative, non-fermenting bacteria and is used in infection and seizure-related studies. The solid form is intended for laboratory preparation and experimental applications.
  • Broad-spectrum aminoglycoside antibiotic.
  • Demonstrated activity against resistant gram-negative non-fermenters.
  • High analytical purity (~98.0%).
  • Water-soluble (H2O 125 mg/mL; requires ultrasonic).
  • Stable when stored sealed and protected from moisture and light.
  • Multiple laboratory pack sizes available for experimental flexibility.
Catalog No. 50-002-13689