Learn More
Medchemexpress LLC Isepamicin sulfate | 67814-76-0 | 98.0% | 679.68 g/mol | C22H43N5O12.xH2SO4 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1005895MG
Encompass_Preferred
Isepamicin sulfate is a broad-spectrum aminoglycoside antibiotic provided as a research reagent. It exhibits activity against Gram-negative, non-fermenting bacteria and is used in studies of antimicrobial effects, vascular regulation, uterine motility, antidiuretic responses, and seizure research. The material is supplied as a sulfate salt with high reported purity for laboratory use and is intended for research purposes only.
  • Broad-spectrum aminoglycoside antibiotic for research applications.
  • Shows activity against Gram-negative non-fermenting bacteria.
  • Used in studies of vascular regulation and uterine motility.
  • Applicable to seizure and antidiuretic research.
  • High purity suitable for laboratory assays.
  • Available in small research pack sizes for experimental use.
Catalog No. 50-002-13718