Learn More
Medchemexpress LLC Isepamicin sulfate | 67814-76-0 | MFCD01683537 | 98.0% | 667.68 g/mol | C22H43N5O12.xH2SO4 | 50 MG

Supplier: Medchemexpress LLC HY10058950MG
Isepamicin sulfate is a research-grade aminoglycoside antibiotic (sulfate salt) with broad-spectrum activity against Gram-negative non-fermenting bacteria, supplied as a white to off-white solid for in vitro and in vivo studies.
- High purity (98.0%) suitable for experimental use.
- Sulfate salt form, provided as a solid with consistent appearance.
- Recommended solvents: water (125 MG/ML) and PBS (100 MG/ML); requires ultrasonic for dissolution.
- Available in multiple package sizes, including 50 MG, for flexible experimental needs.
- Store sealed away from moisture and light; solution storage: -20°C short-term, -80°C long-term.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.