Learn More
Medchemexpress LLC Isepamicin sulfate | 67814-76-0 | MFCD01683537 | 98.0% | 667.7 g/mol | C22H43N5O12 · xH2SO4 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY10058910MG
Encompass_Preferred
Isepamicin sulfate is a research-grade aminoglycoside antibiotic used in antimicrobial and pharmacology studies. It displays activity against certain Gram-negative non-fermenting bacteria and is employed in experimental models investigating vascular effects, uterine contractility, diuresis, and seizure-related responses.
  • High purity (98.0%) suitable for research applications.
  • Solid form, convenient for storage and formulation.
  • Active against Gram-negative non-fermenting bacteria.
  • Useful for studies of vascular blood flow and blood pressure.
  • Applicable to research on uterine contractility and seizure models.
  • Recommended storage: 4°C sealed; in solvent, -80°C for long-term storage.
Catalog No. 50-002-13501