Learn More
Medchemexpress LLC Isethionic acid (80% in water) | 107-36-8 | MFCD00242599 | 80% | 126.13 | 1 ML
SDP
Supplier:  Medchemexpress LLC 10MM/1MLHYY0095
Encompass_Preferred
Isethionic acid is a calcium binder and anionic detergent that enhances mitochondrial calcium binding capacity by competitively binding to calcium binding sites on the outer mitochondrial membrane. It can inhibit calcium-activated mitochondrial respiration. This product can be used to study the mechanism of mitochondrial calcium metabolism, inhibit barnacle larvae, and regulate mitochondrial calcium transport.
  • Enhances mitochondrial calcium binding capacity
  • Inhibits calcium-activated mitochondrial respiration
  • Inhibits barnacle larvae attachment
  • Regulates mitochondrial calcium transport
  • Enhances ATP-dependent calcium uptake
  • Used to study mitochondrial calcium metabolism
  • Relevant to human gut microbiota metabolites
  • Identified as ketones, aldehydes, acids
  • Marine natural product
  • Used in metabolic disease research
  • Endogenous metabolite
Catalog No. 50-003-65191