Learn More
Medchemexpress LLC HY-104012 10mg Medchemexpress, Islatravir CAS:865363-93-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10401210MG
Encompass_Preferred
Medchemexpress, HY-104012 10mg Islatravir CAS:865363-93-5 Islatravir (MK-8591) is a potent anti-HIV-1 agent, acting as a nucleoside reverse transcriptase inhibitor, with EC50s of 0.068 nM, 3.1 nM and 0.15 nM for HIV-1 (WT), HIV-1 (M184V), HIV-1 (MDR), respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8187