Learn More
Selleck Chemical LLC ISO-1 5mg 478336-92-4
SDP
Supplier:  Selleck Chemical LLC S77325MG
Encompass_Preferred
ISO-1 is an inhibitor of MIF (Macrophage migration inhibitory factor) D-dopachrome tautomerase activity with an IC50 of ~7 ?M. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8215