Learn More
Bio Assay Works Iso-Gold™ Rapid Human Antibody Isotyping Kit, 10 tests
Supplier:  Bio Assay Works HISOT010
Encompass
Determining the class and subclass of a human antibody is important in a variety of research areas including infectious disease, cancer, autoimmune diseases, immunodeficiency disorders, and drug and vaccine development. Isotyping is also important when developing and purifying humanized antibodies.The Iso-Gold™ Rapid Human Antibody Isotyping Kit makes it quick and easy to determine human antibody class, subclass and light chain type. Utilizing only two dipsticks, this 5-minute assay provides ELISA-like sensitivity without the time and effort. It can be utilized with cell culture samples and purified antibodies.Specifications:Kit Contents:Human IgG1, IgG2, IgG3 , and IgG4 tests, 10 dipsticksHuman IgA, IgM, κ and λ tests, 10 dipsticksSample Diluent Buffer, 4 mLStorage Temperature: Store Sample Diluent Buffer refrigerated (4-8°C). Dipsticks can be stored at room temperature or 4-8°C.Shipping Temperature: Refrigerated (2-8°C)Stability: Two years from date of manufacture
Catalog No. NC1481181