Learn More
TARGETMOL CHEMICALS INC ISO-RJW100 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T643635MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. (Iso)-RJW100 is a potent liver receptor homolog 1 (LRH-1 NR5A2) and steroidogenic factor-1 (SF-1 NR5A1) agonist with pEC 50 s of 6.4 and 7.2 respectively. purity: 99%
Catalog No. 50-257-9368