Learn More
Medchemexpress LLC Isoalantolactone | 470-17-7 | 232.32 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYN078010MM/1ML
Encompass_Preferred
Isoalantolactone is an apoptosis inducer and alkylating agent that has demonstrated cytotoxic activity against various cancer cell lines. It inhibits cell growth in a dose-dependent manner and is well-tolerated in animal studies, making it suitable for a range of research applications.
  • Apoptosis inducer
  • Acts as an alkylating agent
  • Exhibits cytotoxic activity against K562, PANC-1, BxPC3, and HPAC cells
  • Suitable for both in vitro and in vivo research
  • Well tolerated in CD1 mice at 100 mg/kg
Catalog No. 50-004-16768