Learn More
Medchemexpress LLC Isoalantolactone | 470-17-7 | MFCD08689940 | 232.32 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYN0780
Encompass_Preferred
Isoalantolactone is an apoptosis inducer and alkylating agent. Classified as a terpenoid and sesquiterpene, it is found in Inula helenium L. plants and is considered an endogenous metabolite. It is intended for research and analytical applications.
  • Induces apoptosis
  • Acts as an alkylating agent
  • Classified as a terpenoid and sesquiterpene
  • Derived from Inula helenium L. plants
  • Functions as an endogenous metabolite
  • Suitable for research and analytical applications
Catalog No. 50-003-89855