Learn More
Crescent Chemical Co Inc ISOAMYL ALCOHOL MOL BIO GRADE
SDP
Supplier:  Crescent Chemical Co Inc SE39557.01
Encompass_Preferred
Isoamyl alcohol molecular biology grade DNase, RNase, Proteases not detected. Suitable for use in nucleic acid purification. Isoamyl alcohol prevents foaming during nucleic acid extraction with Phenol:Chloroform:Isoamyl alcohol. Size - 250ML Storage Cond
Catalog No. 50-248-384