Learn More
Medchemexpress LLC Isobutamben | 94-14-4 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB1665100MG
Encompass_Preferred
Isobutamben (Standard) is the analytical standard of Isobutamben (HY-B1665). It is intended for research and analytical applications and is an amino ester-type local anaesthetic agent. This compound is a reference standard supplied assay, commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Analytical standard for Isobutamben
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
  • Amino ester-type local anaesthetic agent
  • For research use only
Catalog No. 50-004-12357