Learn More
Cayman Chemical IsobutyrylLcarnItInchLR 10mg
SDP
Supplier:  Cayman Chemical 3710310 MG
Encompass_Preferred
An acylcarnitine; elevated levels are associated with isobutyryl-CoA dehydrogenase deficiency
Catalog No. 50-291-4968