Learn More
Medchemexpress LLC Isochlorogenic acid A | 2450-53-5 | MFCD00951290 | 99.9% | 516.45 | C25H24O12 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN005610MG
Encompass_Preferred
Isochlorogenic acid A (3,5-dicaffeoylquinic acid) is a natural phenolic compound provided as an analytical reference standard for research and method development. Supplied in a 10 mg laboratory-friendly format, it is used for qualitative and quantitative analyses by HPLC, GC, and MS, and has reported antioxidant, anti-inflammatory, and antiviral activities.
  • Analytical standard suitable for HPLC, GC, and MS.
  • High reported purity for accurate quantitation.
  • Natural phenolic acid with antioxidant and anti-inflammatory properties.
  • Supplied in a small, laboratory-friendly 10 mg format for method development and reference use.
Catalog No. 50-002-96994