Learn More
Medchemexpress LLC Isocitric acid (Standard) | 320-77-4 | 99.9% | 192.12 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY11322850MG
Encompass_Preferred
Isocitric acid (Standard) is an analytical standard and endogenous metabolite intended for research and analytical applications. It is found in saliva and cellular cytoplasm and is used in the study of Alzheimer's Disease, Lewy Body Dementia, and Anoxia. This compound serves as a reference standard for various assays, commonly utilized in qualitative, quantitative, and methodological research involving HPLC, GC, and MS.
  • Analytical standard for research and analytical applications
  • Endogenous metabolite present in saliva and cellular cytoplasm
  • Used in research for Alzheimer's Disease, Lewy Body Dementia, and Anoxia
  • Reference standard for assays
  • Utilized in qualitative, quantitative, and methodological research experiments
  • Compatible with techniques such as HPLC, GC, and MS
Catalog No. 50-004-37252