Learn More
Apexbio Technology LLC Isoconazole nitrate 24168-96-5 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B19565.1
Encompass_Preferred
Isoconazole nitrate (24168-96-5) is a small-molecule inhibitor targeting ergosterol biosynthesis It is designed to inhibit the synthesis of ergosterol a key structural component of fungal cell membranes thereby compromising membrane integrity and biological function Isoconazole nitrate exerts its biological activity primarily through inhibition of ergosterol synthesis In in vitro studies isoconazole nitrate demonstrates antifungal activity by disrupting fungal cell viability and proliferation and also exhibits activity against certain gram-positive bacterial strains Based on these pharmacological properties isoconazole nitrate holds research potential in evaluating antifungal activity (such as MIC or IC50 values) investigating mechanism-related effects and exploring pharmacokinetics tissue distribution and therapeutic efficacy in experimental models of fungal or bacterial infection
Catalog No. 50-000-94609