Learn More
Apexbio Technology LLC Isocorydine 475-67-2 20mg
SDP
Supplier:  Apexbio Technology LLC BA748420
Encompass_Preferred
Isocorydine (CAS 475-67-2) is an isoquinoline alkaloid originally isolated from Menispermum dauricum Studies have shown that isocorydine in combination with doxorubicin (DOX) exhibits potential in targeting hepatocellular carcinoma (HCC) This compound is utilized in research investigating mechanisms of chemosensitization and exploring novel therapeutic strategies for liver cancer particularly in enhancing the efficacy of conventional chemotherapeutic agents
Catalog No. 50-000-88778