Learn More
TARGETMOL CHEMICALS INC Isocorydine hydrochloride 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T429110MG
Encompass_Preferred
Also available in 1 mL, 1 mg, 2 mg, 5 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. (+)-Isocorydine hydrochloride (Isocorydine HCl) is a eukaryote protein kinases Inhibitor. Purity 99.72%
Catalog No. 50-287-8770