Learn More
Medchemexpress LLC HY-N0775 5mg Medchemexpress, Isocorynoxeine CAS:51014-29-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN07755MG
Encompass_Preferred
Medchemexpress, HY-N0775 5mg Isocorynoxeine CAS:51014-29-0 Isocorynoxeine, an isorhynchophylline-related alkaloid, exhibits a dose-dependent inhibition of 5-HT 2A receptor-mediated current response with an IC 50 of 72.4 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3711