Learn More
Medchemexpress LLC Isocytosine | 108-53-2 | 111.10 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0022725G
Encompass_Preferred
Isocytosine is a non-natural nucleobase and an isomer of cytosine. It is used in combination with Isoguanine in studies of unnatural nucleic acid analogues of the normal base pairs in DNA and as a nucleobase of hachimoji RNA.
  • Non-natural nucleobase and an isomer of cytosine.
  • Used in studies of unnatural nucleic acid analogues of normal base pairs in DNA.
  • Used as a nucleobase of hachimoji RNA.
Catalog No. 50-004-45622