Learn More
Medchemexpress LLC Isodeoxycholic acid | 566-17-6 | MFCD00036859 | 99.6% | 392.57 g·mol⁻¹ | C24H40O4 | 10 MG

Supplier: Medchemexpress LLC HYW39929710MG
Isodeoxycholic acid is a bile acid (7α,12α-dihydroxycholanoic acid) and the 3β-epimer of ursodeoxycholic acid. It is provided as a research-grade reagent for biochemical and pharmacological studies of bile acid function, choleresis, and liver biochemistry.
- High reported purity: 99.6%.
- Available in small pack sizes suitable for analytical and exploratory work.
- Molecular formula C24H40O4; molecular weight 392.57 g·mol⁻¹.
- CAS number 566-17-6 for unambiguous identification.
- For research use only; not for human or veterinary use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.