Learn More
TARGETMOL CHEMICALS INC Isodeoxycholic Acid 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T365855MG
Encompass_Preferred
Also available in 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Isodeoxycholic Acid is a bile acid that is formed via epimerization of deoxycholic acid by intestinal bacteria. It has a greater critical micelle concentration than DCA, indicating reduced detergent activity, and is less active than DCA in inhibiting growth in a panel of seven gut commensal bacteria species. Isodeoxycholic Acid(0.1%) inhibits spore germination induced by taurocholic acid in several C. difficile strains, as well as decreases the cytotoxicity of C. difficile culture supernatants to Vero cells. Plasma levels of isodeoxycholic acid are decreased in a rat model of high-fat diet-induced obesity compared with rats fed a normal diet. Purity 98.12%
Catalog No. 50-287-6212