Learn More
Medchemexpress LLC Isodeoxycholic acid (7α,12α-dihydroxycholanoic acid) | 566-17-6 | MFCD00036859 | 99.6% | 392.57 | C24H40O4 | 1 ML

Supplier: Medchemexpress LLC HYW39929710MM/1ML
Isodeoxycholic acid (7α,12α-dihydroxycholanoic acid) is the 3β-epimer of ursodeoxycholic acid supplied as a 10 mM solution in DMSO (1 mL). It is intended for research use and has documented effects on choleresis and liver biochemistry. Refer to the product documentation for handling and safety.
- Supplied as a 10 mM solution in DMSO (1 mL).
- High purity: 99.6%.
- CAS number 566-17-6.
- Molecular formula C24H40O4; molecular weight 392.57.
- Recommend in vitro solvent: DMSO; sonication and freshly opened DMSO advised.
- Suggested in vivo vehicle: 10% DMSO and 90% corn oil (solubility ≥ 2.5 mg/mL).
- Storage: powder at -20°C for up to 3 years; in solvent at -80°C for up to 6 months.
- For research use only; not for human or clinical use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.