Learn More
Medchemexpress LLC Isodeoxycholic acid (7α,12α-dihydroxycholanoic acid) | 566-17-6 | MFCD00036859 | 99.6% | 392.57 g·mol⁻¹ | C24H40O4 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYW3992971MG
Encompass_Preferred
Isodeoxycholic acid (7α,12α-dihydroxycholanoic acid) is a bile acid (CAS 566-17-6, C24H40O4) and the 3β-epimer of ursodeoxycholic acid. Supplied as a high-purity white to off-white solid, it is intended as a research and analytical standard for studies of choleresis and liver biochemistry. Available in small milligram quantities and as ready-to-use DMSO solutions; follow recommended storage conditions.
Catalog No. 50-002-81492