Learn More
Medchemexpress LLC Isodeoxycholic acid (7α,12α-dihydroxycholanoic acid) | 566-17-6 | MFCD00036859 | 99.6% | 392.57 g/mol | C24H40O4 | 25 MG
SDP
Supplier:  Medchemexpress LLC HYW39929725MG
Encompass_Preferred
Isodeoxycholic acid is a bile acid (7α,12α-dihydroxycholanoic acid) and the 3β-epimer of ursodeoxycholic acid. Supplied as a solid analytical standard for research use, it is suitable for biochemical studies, reference standards, and analytical method development. Not for clinical or human use.
  • High purity (99.6%).
  • Solid powder form for analytical use.
  • Suitable as an analytical standard for biochemical assays.
  • Small pack sizes appropriate for laboratory work.
  • Stable when stored as a powder at -20°C or 4°C.
  • Intended for research use only.
Catalog No. 50-002-81153