Learn More
Cayman Chemical IsoferulIc AcId 5g
SDP
Supplier:  Cayman Chemical 227155 G
Encompass_Preferred
A cinnamic acid derivative that has antidiabetic activity; activates α1-adrenergic receptors (IC50 = 1.4 µM) to enhance secretion of β-endorphin (EC50 = 52.2 nM); increases glucose uptake and enhances glycogen synthesis in isolated soleus muscles of streptozocin-induced diabetic rats; induces a dose-dependent reduction in plasma glucose levels in streptozocin-induced diabetic rats; inhibits absorbance of intestinal maltase and sucrose via α-glucosidase in vitro. (IC50s = 760 and 450 µM, respectively)
Catalog No. 50-299-4714