Learn More
Medchemexpress LLC Isoflavone | 574-12-9 | 222.24 | 500 MG
SDP
Supplier:  Medchemexpress LLC HYW006405500MG
Encompass_Preferred
Isoflavone is an orally available bioactive component of soy phytoestrogen with lipid-lowering and antioxidant activities. It prevents various chronic diseases by regulating fatty acid oxidation in the liver and gene expression in adipose tissue. Additionally, isoflavone is valuable in cancer and cardiovascular disease research.
  • Orally available bioactive component of soy phytoestrogen
  • Exhibits lipid-lowering and antioxidant activities
  • Prevents chronic diseases by regulating fatty acid oxidation and gene expression
  • Valuable for cancer and cardiovascular disease research
  • Solid appearance
  • Light brown to brown color
  • Ships at room temperature in continental US
Catalog No. 50-004-47451