Learn More
Beckman Coulter IsoFlow Sheath Fluid, 4 x 1.8 L
Supplier:  Beckman Coulter 8547008
Encompass_Preferred
This isotonic fluid is specifically formulated for use in flow cytometers. IsoFlow Sheath Fluid is carefully manufactured for low particulate and fluorescence backgrounds to ensure superior signal to noise ratio measurements.
Catalog No. NC1939192