Learn More
Medchemexpress LLC Isoformononetin-d3 | 5 MG

Supplier: Medchemexpress LLC HYN7501S5MG
Isoformononetin-d3 is a deuterium labeled version of Isoformononetin, an analog of Daidzein. This compound exhibits immunoprotective effects and can inhibit the differentiation of Th17 and B-cells lymphopoiesis, thereby promoting osteogenesis in estrogen-deficient bone loss conditions.
- Deuterium labeled compound
- Molecular formula: C16H9D3O4
- Molecular weight: 271.28
- Appearance: Off-white to light yellow solid
- Purity (HPLC): 98.64%
- Isotopic enrichment: 99.77%
- Target: Isotope-labeled compounds
- Immunoprotective effects
- Inhibits differentiation of Th17 and B-cells lymphopoiesis
- Promotes osteogenesis in estrogen-deficient bone loss conditions
- Store at 4°C, sealed, away from moisture
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.