Learn More
Cayman Chemical IsogInkgetIn 10mg
SDP
Supplier:  Cayman Chemical 2510410 MG
Encompass_Preferred
A biflavonoid inhibitor of pre-mRNA splicing; reduces the activity of an mRNA-dependent luciferase reporter, shifts the total RNA extract from predominantly mRNA to pre-mRNA, and arrests cell growth in HEK293 cells at 33 μM; suppresses Con A- and LPS-induced mouse lymphocyte proliferation at 10 μM; suppresses PMA- and A23187-induced arachidonic acid release in rat peritoneal macrophages at 10 μM; dose-dependently reduces MMP-9 activity, expression, and mRNA levels in and decreases cell invasion by HT1080 fibrosarcoma cells from 5-20 μM
Catalog No. 50-293-5423