Learn More
STA PHARMACEUTICAL US LLC isoGNA C(iBu) amidite | 1 g | CAS 1443744-42-0 | InChIKey XGLKXRATAYJCHB-XYMJDGAWSA-N
Supplier: STA PHARMACEUTICAL US LLC STAAM10629KA301GM
isoGNA C(iBu) amidite is a Amidite reagent (Subcategory: isoGNA Series) sold by WuXi TIDES. Offered in 1 g. Store at -20 °C. Ships on dry ice. SDS available for reference.
Specifications
- CAS: 1443744-42-0
- MDL: No data
- InChIKey: XGLKXRATAYJCHB-XYMJDGAWSA-N
- Molecular Weight: 757.868761998
- Molecular Formula: C41H52N5O7P
- Purity: ≥98%
- Container Type: 30 mL Glass (20-400)
- Pack Size: 1 g
- Net Weight: 1 g
- Gross Weight: 47.3 g
- Commodity Code: 29335995
- Country Of Origin: China
- IUPAC: (S)-3-(bis(4-methoxyphenyl)(phenyl)methoxy)-2-(4-isobutyramido-2-oxopyrimidin-1(2H)-yl)propyl (2-cyanoethyl) diisopropylphosphoramidite
- SMILES: O=C1N(C=CC(NC(C(C)C)=O)=N1)[C@@H](COC(C2=CC=C(C=C2)OC)(C3=CC=C(C=C3)OC)C4=CC=CC=C4)COP(N(C(C)C)C(C)C)OCCC#N
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.